Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCHCR1 SIRNA Assays ›
Gene Symbol:
CCHCR1
Gene Name:
coiled-coil alpha-helical rod protein 1
Gene Aliases:
C6orf18, HCR, SBP
Chromosome Location:
Chr.6: 31142439 - 31158238 on Build GRCh38
UniGene ID:
Hs.485075
Species:
Human
siRNA ID
HSS182548
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCHCR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105563.1
NP_001099033.1
17
2515
NM_001105564.1
NP_001099034.1
17
2623
NM_019052.3
NP_061925.2
17
2290
XM_011514702.2
XP_011513004.1
17
2549
XM_011514703.1
XP_011513005.1
18
2362
XM_011514704.2
XP_011513006.1
17
2151
XM_011514705.1
XP_011513007.1
17
2260
XM_017010961.1
XP_016866450.1
17
2657
XM_017010962.1
XP_016866451.1
18
2409
XM_017010963.1
XP_016866452.1
18
2523
XM_017010964.1
XP_016866453.1
17
2287
XM_017010965.1
XP_016866454.1
17
2349
XM_017010966.1
XP_016866455.1
17
2370
XM_017010967.1
XP_016866456.1
17
2473
XM_017010968.1
XP_016866457.1
17
2242
XM_017010969.1
XP_016866458.1
17
2149
XM_017010970.1
XP_016866459.1
16
1935
GenBank mRNA
AB029331.1
-
15
2111
AB112474.1
-
17
2246
AB112475.1
-
17
2597
AF216493.1
-
15
2111
AK000204.1
-
17
2246
AK000217.1
-
17
2247
AK000533.1
-
12
1362
AK289446.1
-
17
2238
AY029160.1
-
16
2167
BC110534.1
-
17
2333
40035162
BC110535.1
-
17
2446
40035164
CR457227.1
-
16
2167
Back To Top
More Information
Gene Ontology Categories:
Function(s)
protein export from nucleus
multicellular organism development
cell differentiation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0