Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019058.2 | NP_061931.1 | 2 | 216 | ||
GenBank mRNA | AF335324.1 | - | 2 | 232 | ||
AK000507.1 | - | 2 | 211 | |||
AL136668.1 | - | 2 | 215 | |||
AY090097.1 | - | 2 | 132 | |||
BC000708.1 | - | 2 | 122 | 3507244 | ||
BC007714.1 | - | 2 | 204 | 4302878 | ||
BC015236.1 | - | 2 | 190 | 3906374 |