Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021129.3 | NP_066952.1 | 6 | 600 | ||
GenBank mRNA | AB026723.1 | - | 6 | 523 | ||
AF092439.1 | - | 6 | 570 | |||
AF108211.1 | - | 6 | 481 | |||
AF119665.1 | - | 6 | 570 | |||
AF154065.1 | - | 6 | 538 | |||
AF217186.1 | - | 6 | 493 | |||
AK058193.1 | - | 6 | 593 | |||
BC001022.2 | - | 6 | 575 | 3345496 | ||
BC061581.1 | - | 6 | 583 | 6526628 | ||
BC105034.1 | - | 6 | 566 | 8144037 | ||
BC105036.1 | - | 6 | 566 | 8144039 | ||
BC107882.1 | - | 6 | 534 | 6495107 | ||
BU520836.1 | - | 6 | 581 | |||
EU794626.1 | - | 6 | 600 |