Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307977.1 | NP_001294906.1 | 7 | 751 | ||
NM_018093.3 | NP_060563.2 | 8 | 830 | |||
XM_005274055.2 | XP_005274112.1 | 8 | 2691 | |||
GenBank mRNA | AK001301.1 | - | 8 | 830 | ||
AK025383.1 | - | 3 | 265 | |||
AK292330.1 | - | 8 | 1200 | |||
AY500828.1 | - | 7 | 682 | |||
BC006351.2 | - | 7 | 673 | 4111690 | ||
BU740579.1 | - | 5 | 557 |