Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018364.4 | NP_060834.2 | 7 | 2186 | ||
NR_130896.1 | 8 | 2304 | ||||
GenBank mRNA | AK002082.1 | - | 7 | 2001 | ||
AK022166.1 | - | 6 | 1365 | |||
AK292552.1 | - | 7 | 2186 | |||
BC026155.1 | - | 8 | 2126 | 4827655 | ||
CR627402.1 | - | 8 | 1779 |