Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017824.4 | NP_060294.1 | 5 | 975 | ||
XM_011539886.2 | XP_011538188.1 | 6 | 1123 | |||
GenBank mRNA | AB191202.1 | - | 5 | 731 | ||
AK000452.1 | - | 5 | 977 | |||
AK294799.1 | - | 5 | 1049 | |||
AK309175.1 | - | 4 | 844 | |||
BC015480.1 | - | 5 | 935 | 3905766 |