Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019098.4 | NP_061971.3 | 10 | 1121 | ||
XM_011517138.2 | XP_011515440.1 | 8 | 776 | |||
GenBank mRNA | AF228520.1 | - | 7 | 734 | ||
AF272900.1 | - | 10 | 1119 | |||
BC150601.1 | - | 10 | 1099 | 9056971 |