Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017520.3 | NP_059990.2 | 10 | 2173 | ||
XM_011535124.2 | XP_011533426.1 | 10 | 2174 | |||
GenBank mRNA | AK056785.1 | - | 3 | 220 | ||
AK300258.1 | - | 10 | 2173 | |||
AL832864.1 | - | 10 | 2122 | |||
BC003542.1 | - | 10 | 2081 | 3611719 | ||
BC046214.1 | - | 4 | 360 |