Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316936.1 | NP_001303865.1 | 2 | 382 | ||
NM_001316937.1 | NP_001303866.1 | 3 | 455 | |||
NM_001316938.1 | NP_001303867.1 | 4 | 525 | |||
NM_001316939.1 | NP_001303868.1 | 4 | 499 | |||
NM_001316940.1 | NP_001303869.1 | 3 | 377 | |||
NM_001316941.1 | NP_001303870.1 | 2 | 408 | |||
NM_001316942.1 | NP_001303871.1 | 4 | 553 | |||
NM_017703.2 | NP_060173.1 | 3 | 481 | |||
XM_006722782.3 | XP_006722845.2 | 2 | 232 | |||
XM_017026912.1 | XP_016882401.1 | 3 | 357 | |||
GenBank mRNA | AA524458.1 | - | 2 | 124 | ||
AK000195.1 | - | 3 | 433 | |||
AK027004.1 | - | 4 | 501 | |||
AK093760.1 | - | 3 | 428 | |||
AL389936.1 | - | 2 | 216 | |||
BC001586.2 | - | 3 | 385 | 2966439 | ||
BI760557.1 | - | 4 | 235 | |||
BM019575.1 | - | 4 | 475 | |||
BP367273.1 | - | 3 | 350 | |||
BX647073.1 | - | 2 | 318 | |||
DB127388.1 | - | 3 | 455 |