Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017708.3 | NP_060178.2 | 3, 4 | 811 | ||
XM_005259019.1 | XP_005259076.1 | 4, 5 | 2135 | |||
XM_011527049.2 | XP_011525351.1 | 4, 5 | 2120 | |||
GenBank mRNA | AK000207.1 | - | 3, 4 | 937 | ||
BC111970.1 | - | 3, 4 | 811 | 8327438 | ||
BC111972.1 | - | 3, 4 | 811 | 8327440 |