Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017802.3 | NP_060272.3 | 11, 12 | 2253 | ||
NR_075098.1 | 11, 12 | 2191 | ||||
GenBank mRNA | AK056233.1 | - | 11, 12 | 1638 | ||
AK098430.1 | - | 5, 6 | 773 | |||
BC047240.1 | - | 11, 12 | 2253 | 5274510 | ||
BC094840.1 | - | 4, 5 | 1009 |