Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017802.3 | NP_060272.3 | 2 | 693 | ||
NR_075098.1 | 2 | 631 | ||||
GenBank mRNA | AK056233.1 | - | 2 | 78 | ||
BC047240.1 | - | 2 | 693 | 5274510 | ||
BM743055.1 | - | 2 | 495 |