Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142317.1 | NP_001135789.1 | 4 | 567 | ||
NM_001142318.1 | NP_001135790.1 | 4 | 465 | |||
NM_001324355.1 | NP_001311284.1 | 5 | 716 | |||
NM_017853.2 | NP_060323.1 | 4 | 616 | |||
GenBank mRNA | AK000518.1 | - | 4 | 591 | ||
AK222889.1 | - | 4 | 583 | |||
AY566808.1 | - | 3 | 305 | |||
BC009646.1 | - | 4 | 557 | 3458545 | ||
BM849401.1 | - | 5 | 617 | |||
CD517462.1 | - | 4 | 513 | |||
CR457240.1 | - | 3 | 305 |