Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017857.3 | NP_060327.3 | 13, 14 | 1857 | ||
XM_017017942.1 | XP_016873431.1 | 13, 14 | 1896 | |||
GenBank mRNA | AB072360.3 | - | 13, 14 | 1581 | ||
AB099291.1 | - | 13, 14 | 1679 | |||
AK000522.1 | - | 9, 10 | 1030 | |||
AK001790.1 | - | 6, 7 | 822 | |||
AK074432.1 | - | 13, 14 | 1775 | |||
AK094226.1 | - | 12, 13 | 1886 | |||
BC004176.2 | - | 6, 7 | 861 | |||
BC004210.2 | - | 6, 7 | 861 | |||
BC007709.1 | - | 13, 14 | 1736 | 4053628 |