Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166304.1 | NP_001159776.1 | 6 | 812 | ||
NM_017861.3 | NP_060331.3 | 5 | 758 | |||
GenBank mRNA | AK000529.1 | - | 6 | 775 | ||
AK294340.1 | - | 7 | 1112 | |||
CN404010.1 | - | 3 | 434 |