Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM132A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017870.3 | NP_060340.2 | 2 | 413 | ||
NM_178031.2 | NP_821174.1 | 2 | 413 | |||
XM_017017951.1 | XP_016873440.1 | 1 | 2010 | |||
XM_017017952.1 | XP_016873441.1 | 1 | 2009 | |||
GenBank mRNA | AB046803.1 | - | 1 | 256 | ||
AL834133.1 | - | 1 | 275 | |||
BC028106.1 | - | 2 | 612 | |||
BC032059.1 | - | 2 | 373 | |||
BC114569.1 | - | 2 | 378 | 40036200 |