Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017875.2 | NP_060345.2 | 1, 2 | 458 | ||
XM_017006710.1 | XP_016862199.1 | 1, 2 | 457 | |||
GenBank mRNA | AK000558.1 | - | 1, 2 | 434 | ||
AK225388.1 | - | 1, 2 | 410 | |||
BC013194.1 | - | 1, 2 | 119 | 3918744 | ||
CR457242.1 | - | 1, 2 | 57 | |||
DB113609.1 | - | 1, 2 | 458 |