Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF159 SIRNA Assays ›
Gene Symbol:
C1orf159
Gene Name:
chromosome 1 open reading frame 159
Gene Aliases:
-
Chromosome Location:
Chr.1: 1081811 - 1116356 on Build GRCh38
UniGene ID:
Hs.235095
Species:
Human
siRNA ID
HSS182779
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf159
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017891.4
NP_060361.4
7
819
XM_005244765.2
XP_005244822.1
7
557
XM_006710717.2
XP_006710780.1
8
913
XM_006710718.2
XP_006710781.1
7
841
XM_011541651.2
XP_011539953.1
8
828
XM_011541652.2
XP_011539954.1
10
1825
XM_011541653.2
XP_011539955.1
9
922
XM_011541655.2
XP_011539957.1
8
1866
XM_011541656.2
XP_011539958.1
8
1396
XM_011541658.2
XP_011539960.1
8
1583
XM_011541659.2
XP_011539961.1
8
697
XM_011541660.2
XP_011539962.1
8
1232
XM_011541661.2
XP_011539963.1
8
678
XM_011541662.2
XP_011539964.1
9
1021
XM_011541663.2
XP_011539965.1
9
706
XM_011541665.2
XP_011539967.1
8
598
XM_017001572.1
XP_016857061.1
9
2012
XM_017001573.1
XP_016857062.1
8
1714
XM_017001574.1
XP_016857063.1
8
1918
XM_017001575.1
XP_016857064.1
9
876
XM_017001576.1
XP_016857065.1
9
1968
XM_017001577.1
XP_016857066.1
7
785
XM_017001578.1
XP_016857067.1
7
1244
XM_017001579.1
XP_016857068.1
7
557
XM_017001580.1
XP_016857069.1
9
2012
GenBank mRNA
AK000591.1
-
7
554
AK054793.1
-
6
1654
AK057368.1
-
9
706
AK124054.1
-
8
649
AK128271.1
-
2
575
AK128434.1
-
7
1498
AK301998.1
-
6
525
AY358490.1
-
7
558
BC008788.2
-
6
495
3627081
BX394177.2
-
7
704
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline