Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM255A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104544.1 | NP_001098014.1 | 9 | 1128 | ||
NM_001104545.1 | NP_001098015.1 | 7 | 876 | |||
NM_017938.3 | NP_060408.3 | 10 | 1200 | |||
GenBank mRNA | AF450484.1 | - | 10 | 1130 | ||
AK000723.1 | - | 5 | 583 | |||
AK056403.1 | - | 9 | 1131 | |||
AK289684.1 | - | 10 | 1091 | |||
BC047054.1 | - | 10 | 1097 | 5313196 |