Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017946.3 | NP_060416.1 | 1 | 400 | ||
NR_046478.1 | 1 | 400 | ||||
NR_046479.1 | 1 | 400 | ||||
GenBank mRNA | AK000738.1 | - | 1 | 252 | ||
AK309552.1 | - | 1 | 261 | |||
AY358643.1 | - | 1 | 280 | |||
BC005206.1 | - | 1 | 283 | 4042173 | ||
DA166879.1 | - | 1 | 401 | |||
DB498479.1 | - | 1 | 20 | |||
JQ797475.1 | - | 1 | 107 |