Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256394.1 | NP_001243323.1 | 8 | 2582 | ||
NM_017948.5 | NP_060418.4 | 7 | 2478 | |||
NR_046106.1 | 7 | 2478 | ||||
XM_006717166.3 | XP_006717229.1 | 7 | 2344 | |||
XM_006717167.3 | XP_006717230.1 | 7 | 2466 | |||
XM_006717168.3 | XP_006717231.1 | 7 | 2237 | |||
XM_006717169.3 | XP_006717232.1 | 8 | 2497 | |||
XM_006717170.3 | XP_006717233.1 | 8 | 2365 | |||
XM_006717172.3 | XP_006717235.1 | 8 | 2689 | |||
XM_006717173.3 | XP_006717236.1 | 9 | 2802 | |||
XM_011518824.2 | XP_011517126.1 | 6 | 2062 | |||
XM_011518827.2 | XP_011517129.1 | 5 | 2106 | |||
XM_017014876.1 | XP_016870365.1 | 7 | 2344 | |||
XM_017014877.1 | XP_016870366.1 | 8 | 2365 | |||
XM_017014878.1 | XP_016870367.1 | 8 | 2815 | |||
XM_017014879.1 | XP_016870368.1 | 8 | 2644 | |||
XM_017014880.1 | XP_016870369.1 | 8 | 2444 | |||
GenBank mRNA | AB105104.1 | - | 7 | 2444 | ||
AB109030.1 | - | 6 | 2141 | |||
AK000743.1 | - | 1 | 519 | |||
AK022755.1 | - | 4 | 1891 | |||
AK024245.1 | - | 1 | 1236 | |||
AK225322.1 | - | 1 | 519 | |||
AK297355.1 | - | 8 | 2289 | |||
AK307408.1 | - | 7 | 2369 | |||
BC013788.1 | - | 1 | 726 | |||
BC064942.1 | - | 1 | 1401 | |||
BC146810.1 | - | 8 | 2338 | 8860369 | ||
CR627363.1 | - | 7 | 2222 |