Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256394.1 | NP_001243323.1 | 6 | 810 | ||
NM_017948.5 | NP_060418.4 | 5 | 706 | |||
NR_046106.1 | 5 | 706 | ||||
XM_006717166.3 | XP_006717229.1 | 5 | 572 | |||
XM_006717167.3 | XP_006717230.1 | 5 | 694 | |||
XM_006717168.3 | XP_006717231.1 | 5 | 465 | |||
XM_006717169.3 | XP_006717232.1 | 6 | 725 | |||
XM_006717170.3 | XP_006717233.1 | 6 | 593 | |||
XM_006717172.3 | XP_006717235.1 | 6 | 917 | |||
XM_006717173.3 | XP_006717236.1 | 7 | 1030 | |||
XM_011518824.2 | XP_011517126.1 | 4 | 290 | |||
XM_011518827.2 | XP_011517129.1 | 3 | 334 | |||
XM_017014876.1 | XP_016870365.1 | 5 | 572 | |||
XM_017014877.1 | XP_016870366.1 | 6 | 593 | |||
XM_017014878.1 | XP_016870367.1 | 6 | 1043 | |||
XM_017014879.1 | XP_016870368.1 | 6 | 872 | |||
XM_017014880.1 | XP_016870369.1 | 6 | 672 | |||
GenBank mRNA | AB105104.1 | - | 5 | 672 | ||
AB109030.1 | - | 4 | 369 | |||
AK001049.1 | - | 5 | 588 | |||
AK022755.1 | - | 2 | 119 | |||
AK297355.1 | - | 5 | 589 | |||
AK307408.1 | - | 5 | 597 | |||
BC146810.1 | - | 6 | 566 | 8860369 | ||
CR627363.1 | - | 5 | 450 |