Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291504.1 | NP_001278433.1 | 1 | 437 | ||
NM_001291505.1 | NP_001278434.1 | 1 | 437 | |||
NM_001316325.1 | NP_001303254.1 | 1 | 437 | |||
NM_006241.7 | NP_006232.1 | 1 | 437 | |||
NR_133014.1 | 1 | 437 | ||||
XM_006713682.3 | XP_006713745.1 | 1 | 384 | |||
GenBank mRNA | AJ133812.1 | - | 1 | 64 | ||
AY063767.1 | - | 1 | 63 | |||
BC007655.2 | - | 1 | 286 | 3346573 | ||
BC066921.1 | - | 1 | 422 | |||
BI549862.1 | - | 1 | 356 | |||
BI560631.1 | - | 1 | 200 | |||
X78873.1 | - | 1 | 64 | |||
Z29646.1 | - | 1 | 61 |
Gene Symbol: PPP1R2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038443.1 | 1 | 86 | |||
GenBank mRNA | BC066922.1 | - | 1 | 86 | ||
JF438008.1 | - | 1 | 138 |