Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039775.3 | NP_001034864.2 | 16 | 4566 | ||
XM_005245918.2 | XP_005245975.1 | 16 | 4552 | |||
XM_011541672.1 | XP_011539974.1 | 15 | 4516 | |||
XM_011541673.2 | XP_011539975.1 | 16 | 4791 | |||
GenBank mRNA | AK095339.1 | - | 15 | 1610 | ||
BC136870.1 | - | 12 | 1308 | 9020861 |