Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018009.4 | NP_060479.3 | 6 | 1460 | ||
XM_005253700.3 | XP_005253757.1 | 6 | 1460 | |||
XM_017019545.1 | XP_016875034.1 | 6 | 1608 | |||
XM_017019546.1 | XP_016875035.1 | 6 | 1288 | |||
GenBank mRNA | AI184901.1 | - | 2 | 363 | ||
AK001005.1 | - | 6 | 1364 | |||
AK002056.2 | - | 6 | 1348 | |||
AK098035.1 | - | 7 | 1422 | |||
AK222956.1 | - | 6 | 1348 | |||
AK223163.1 | - | 6 | 1378 | |||
BC015017.2 | - | 6 | 1387 | 3916213 |