Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321829.1 | NP_001308758.1 | 4 | 1124 | ||
NM_018017.3 | NP_060487.2 | 3 | 999 | |||
NR_135815.1 | 4 | 1124 | ||||
XM_011539915.2 | XP_011538217.1 | 2 | 342 | |||
XM_011539916.2 | XP_011538218.1 | 3 | 1091 | |||
GenBank mRNA | AF273054.1 | - | 2 | 514 | ||
AK307517.1 | - | 3 | 976 | |||
AL834368.1 | - | 2 | 198 | |||
BC100670.1 | - | 4 | 1051 | |||
BC103499.1 | - | 3 | 926 | 40001826 | ||
BC105632.1 | - | 3 | 926 | 40001824 | ||
BC107430.1 | - | 3 | 959 | |||
BC152306.1 | - | 3 | 930 | 40012131 | ||
BX537964.1 | - | 3 | 971 |