Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286247.1 | NP_001273176.1 | 2 | 393 | ||
NM_152663.4 | NP_689876.2 | 2 | 393 | |||
XM_006711410.3 | XP_006711473.1 | 2 | 306 | |||
XM_011509688.1 | XP_011507990.1 | 2 | 307 | |||
XM_017001591.1 | XP_016857080.1 | 2 | 267 | |||
GenBank mRNA | AK001106.1 | - | 2 | 299 | ||
AK301753.1 | - | 2 | 299 | |||
BC047391.1 | - | 2 | 378 | 5298449 |