Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018046.4 | NP_060516.2 | 4, 5 | 1052 | ||
GenBank mRNA | AW958325.1 | - | 4, 5 | 534 | ||
AY500994.1 | - | 4, 5 | 971 | |||
BC032844.1 | - | 4, 5 | 1038 | 5266978 | ||
BC073864.1 | - | 4, 5 | 579 | |||
BE077189.1 | - | 2, 3 | 327 |