Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006634.2 | NP_001006635.1 | 14 | 1339 | ||
NM_018054.5 | NP_060524.4 | 14 | 1339 | |||
XM_011545873.2 | XP_011544175.1 | 15 | 1428 | |||
XM_011545874.2 | XP_011544176.1 | 16 | 1510 | |||
XM_011545875.2 | XP_011544177.1 | 15 | 1428 | |||
XM_011545876.2 | XP_011544178.1 | 15 | 1428 | |||
XM_011545877.2 | XP_011544179.1 | 15 | 1366 | |||
XM_011545878.2 | XP_011544180.1 | 15 | 1428 | |||
XM_017023390.1 | XP_016878879.1 | 15 | 1428 | |||
GenBank mRNA | AJ306731.1 | - | 14 | 1222 | ||
AK023281.1 | - | 11 | 991 | |||
AK125358.1 | - | 5 | 3305 | |||
AK291691.1 | - | 14 | 1292 | |||
AK301500.1 | - | 12 | 1028 | |||
BC080195.1 | - | 14 | 1292 | 6172310 |