Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204831.1 | NP_001191760.1 | 3 | 389 | ||
NM_001204834.1 | NP_001191763.1 | 3 | 389 | |||
NM_018075.3 | NP_060545.3 | 3 | 389 | |||
XM_011533885.2 | XP_011532187.2 | 3 | 355 | |||
XM_011533887.2 | XP_011532189.1 | 3 | 355 | |||
XM_011533889.2 | XP_011532191.1 | 3 | 355 | |||
XM_011533890.2 | XP_011532192.1 | 3 | 355 | |||
XM_017006716.1 | XP_016862205.1 | 3 | 355 | |||
XM_017006717.1 | XP_016862206.1 | 3 | 307 | |||
XM_017006718.1 | XP_016862207.1 | 3 | 334 | |||
XM_017006720.1 | XP_016862209.1 | 3 | 334 | |||
GenBank mRNA | AK096302.1 | - | 3 | 389 | ||
AK292368.1 | - | 3 | 274 | |||
AK297949.1 | - | 3 | 332 | |||
AL832508.1 | - | 3 | 256 | |||
BC038855.1 | - | 3 | 287 |