Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321825.1 | NP_001308754.1 | 2 | 725 | ||
NM_018086.3 | NP_060556.2 | 3 | 895 | |||
XM_011511389.2 | XP_011509691.1 | 4 | 1299 | |||
XM_017004419.1 | XP_016859908.1 | 2 | 10832 | |||
GenBank mRNA | AK001267.1 | - | 1 | 382 | ||
AK125324.1 | - | 3 | 896 | |||
BX649105.1 | - | 3 | 893 |