Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018109.3 | NP_060579.3 | 5 | 1040 | ||
GenBank mRNA | AB194709.1 | - | 5 | 977 | ||
AK001348.1 | - | 5 | 981 | |||
AK022188.1 | - | 5 | 1020 | |||
AY364242.1 | - | 5 | 977 | |||
BC061703.1 | - | 5 | 1035 | 6712967 |