Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127182.1 | NP_001120654.1 | 3 | 603 | ||
NM_018131.4 | NP_060601.3 | 3 | 725 | |||
XM_011539918.1 | XP_011538220.1 | 3 | 739 | |||
XM_011539919.1 | XP_011538221.1 | 3 | 607 | |||
XM_011539920.2 | XP_011538222.1 | 3 | 433 | |||
XM_017016372.1 | XP_016871861.1 | 3 | 579 | |||
XM_017016373.1 | XP_016871862.1 | 3 | 601 | |||
GenBank mRNA | AB091343.1 | - | 3 | 714 | ||
AK001402.1 | - | 2 | 325 | |||
AK223495.1 | - | 3 | 706 | |||
AK315536.1 | - | 3 | 722 | |||
AY788918.1 | - | 3 | 725 | |||
BC008947.2 | - | 3 | 705 | 3028566 | ||
BX640718.1 | - | 4 | 645 |