Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018132.3 | NP_060602.2 | 7 | 605 | ||
XM_005249205.1 | XP_005249262.1 | 7 | 666 | |||
XM_011514723.2 | XP_011513025.2 | 6 | 541 | |||
GenBank mRNA | AK001407.1 | - | 2 | 155 | ||
AK293016.1 | - | 7 | 610 | |||
BC016279.1 | - | 7 | 595 | 3908895 | ||
BI768787.1 | - | 7 | 625 |