Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134415.1 | NP_001127887.1 | 2 | 564 | ||
NM_018138.4 | NP_060608.1 | 2 | 215 | |||
GenBank mRNA | AK001422.1 | - | 2 | 188 | ||
BC025748.1 | - | 2 | 201 | 5200082 | ||
DB023715.1 | - | 2 | 213 | |||
HY017180.1 | - | 2 | 215 |