Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083908.1 | NP_001077377.1 | 1 | 1838 | ||
NM_018139.2 | NP_060609.2 | 1 | 1838 | |||
GenBank mRNA | AK001425.1 | - | 1 | 625 | ||
BC011400.1 | - | 1 | 602 | |||
BC013322.1 | - | 1 | 551 | |||
BX248264.1 | - | 1 | 574 | |||
FJ158843.1 | - | 1 | 1757 |