Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104647.1 | NP_001098117.1 | 7 | 1127 | ||
NM_018155.2 | NP_060625.2 | 7 | 1124 | |||
XM_005247575.4 | XP_005247632.1 | 4 | 2238 | |||
GenBank mRNA | AK001480.1 | - | 7 | 1098 | ||
AK296785.1 | - | 6 | 968 | |||
AL049246.1 | - | 1 | 130 | |||
BC014064.1 | - | 7 | 1060 | 3842022 | ||
BC019859.1 | - | 8 | 1089 | |||
BC036272.1 | - | 8 | 1089 |