Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104647.1 | NP_001098117.1 | 3, 4 | 527 | ||
NM_018155.2 | NP_060625.2 | 3, 4 | 527 | |||
GenBank mRNA | AK001480.1 | - | 3, 4 | 501 | ||
AL136803.1 | - | 3, 4 | 460 | |||
BC014064.1 | - | 3, 4 | 460 | 3842022 | ||
BC019859.1 | - | 3, 4 | 452 | |||
BC036272.1 | - | 3, 4 | 452 |