Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
siRNA ID
HSS182923
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
17
2311
XM_005265279.4
XP_005265336.1
19
2584
XM_005265280.3
XP_005265337.1
18
2401
XM_005265282.3
XP_005265339.1
19
2543
XM_005265283.3
XP_005265340.1
18
2398
XM_011533900.2
XP_011532202.1
21
2750
XM_011533902.2
XP_011532204.1
20
2682
XM_011533903.2
XP_011532205.1
20
2687
XM_017006725.1
XP_016862214.1
20
2657
XM_017006726.1
XP_016862215.1
21
2767
XM_017006727.1
XP_016862216.1
21
2712
XM_017006728.1
XP_016862217.1
20
2708
XM_017006729.1
XP_016862218.1
20
3116
XM_017006730.1
XP_016862219.1
21
2703
XM_017006731.1
XP_016862220.1
20
2686
XM_017006732.1
XP_016862221.1
20
2682
XM_017006733.1
XP_016862222.1
20
3076
XM_017006734.1
XP_016862223.1
20
2649
XM_017006735.1
XP_016862224.1
19
3070
XM_017006736.1
XP_016862225.1
19
3071
XM_017006737.1
XP_016862226.1
19
2990
XM_017006738.1
XP_016862227.1
19
2611
XM_017006739.1
XP_016862228.1
19
2987
XM_017006740.1
XP_016862229.1
19
2528
XM_017006741.1
XP_016862230.1
19
2619
XM_017006742.1
XP_016862231.1
19
2642
XM_017006743.1
XP_016862232.1
20
3116
XM_017006744.1
XP_016862233.1
19
2608
XM_017006745.1
XP_016862234.1
19
2962
XM_017006746.1
XP_016862235.1
19
2959
XM_017006747.1
XP_016862236.1
19
3021
XM_017006748.1
XP_016862237.1
18
2917
XM_017006749.1
XP_016862238.1
18
2483
XM_017006750.1
XP_016862239.1
17
2371
XM_017006751.1
XP_016862240.1
19
2619
XM_017006752.1
XP_016862241.1
20
3116
XM_017006753.1
XP_016862242.1
20
3115
XM_017006754.1
XP_016862243.1
20
2682
XM_017006755.1
XP_016862244.1
19
2619
XM_017006756.1
XP_016862245.1
19
2619
XM_017006757.1
XP_016862246.1
17
2342
XM_017006758.1
XP_016862247.1
17
2473
XM_017006759.1
XP_016862248.1
19
2619
XM_017006760.1
XP_016862249.1
20
2682
XM_017006761.1
XP_016862250.1
20
2691
XM_017006762.1
XP_016862251.1
20
2682
XM_017006763.1
XP_016862252.1
21
2741
XM_017006764.1
XP_016862253.1
19
2619
XM_017006765.1
XP_016862254.1
18
2498
XM_017006766.1
XP_016862255.1
19
2626
GenBank mRNA
AF177387.1
-
16
2200
AF225870.1
-
15
2104
AF225871.1
-
16
2200
AF225872.1
-
16
2200
BC070273.1
-
2
322
BC115009.1
-
16
2215
7962003
BC115011.1
-
16
2216
7962010
BC129934.1
-
17
2245
40082629
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-2ddqc:80/100.66.79.182:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline