Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLPH3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018178.5 | NP_060648.2 | 5 | 1056 | ||
XM_006711428.1 | XP_006711491.1 | 5 | 983 | |||
GenBank mRNA | AJ296153.1 | - | 5 | 884 | ||
AK001549.1 | - | 4 | 684 | |||
AK300635.1 | - | 4 | 882 | |||
AK315594.1 | - | 5 | 1023 | |||
BC013870.2 | - | 4 | 855 | 3837466 |