Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082969.1 | NP_001076438.1 | 6 | 958 | ||
NM_001082970.1 | NP_001076439.1 | 6 | 1513 | |||
NM_001301017.1 | NP_001287946.1 | 6 | 955 | |||
NM_001301019.1 | NP_001287948.1 | 6 | 999 | |||
NM_001301021.1 | NP_001287950.1 | 6 | 996 | |||
NM_018195.3 | NP_060665.3 | 6 | 1516 | |||
NR_103469.1 | 6 | 1557 | ||||
GenBank mRNA | AB096251.1 | - | 6 | 862 | ||
AK001588.1 | - | 3 | 575 | |||
AK125340.1 | - | 6 | 1469 | |||
BC005403.1 | - | 4 | 755 | |||
BC030546.1 | - | 4 | 760 | |||
BC048133.1 | - | 6 | 1444 | 5295844 | ||
BC072455.1 | - | 6 | 880 | 6185048 | ||
BX538107.1 | - | 6 | 1515 |