Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278211.1 | NP_001265140.1 | 2 | 262 | ||
NM_001278212.1 | NP_001265141.1 | 2 | 207 | |||
NM_001278213.1 | NP_001265142.1 | 2 | 156 | |||
NM_018205.3 | NP_060675.1 | 2 | 207 | |||
NM_018239.3 | NP_060709.2 | 2 | 207 | |||
NM_207119.2 | NP_997002.1 | 2 | 520 | |||
XM_017016374.1 | XP_016871863.1 | 2 | 202 | |||
XM_017016375.1 | XP_016871864.1 | 2 | 197 | |||
GenBank mRNA | AK001613.1 | - | 2 | 183 | ||
AK001706.1 | - | 2 | 183 | |||
AK094734.1 | - | 2 | 203 | |||
AY358318.1 | - | 2 | 264 | |||
BC024001.2 | - | 2 | 198 | 4822655 | ||
BG771737.1 | - | 2 | 523 | |||
BM788766.1 | - | 2 | 181 | |||
CF552977.1 | - | 2 | 176 | |||
DA704994.1 | - | 2 | 156 | |||
DA775185.1 | - | 2 | 183 | |||
DA821329.1 | - | 2 | 207 |