Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278211.1 | NP_001265140.1 | 5 | 676 | ||
NM_001278212.1 | NP_001265141.1 | 5 | 621 | |||
NM_001278213.1 | NP_001265142.1 | 4 | 420 | |||
NM_001278214.1 | NP_001265143.1 | 4 | 476 | |||
NM_018205.3 | NP_060675.1 | 4 | 453 | |||
NM_018239.3 | NP_060709.2 | 4 | 471 | |||
NM_207119.2 | NP_997002.1 | 5 | 934 | |||
NR_103467.1 | 3 | 326 | ||||
NR_103468.1 | 3 | 308 | ||||
XM_017016374.1 | XP_016871863.1 | 5 | 616 | |||
XM_017016375.1 | XP_016871864.1 | 5 | 611 | |||
GenBank mRNA | AK001613.1 | - | 4 | 429 | ||
AK001706.1 | - | 4 | 447 | |||
AK094734.1 | - | 5 | 617 | |||
AL122117.1 | - | 2 | 158 | |||
AY358318.1 | - | 5 | 678 | |||
BC024001.2 | - | 5 | 612 | 4822655 | ||
BM788766.1 | - | 5 | 595 | |||
CF552977.1 | - | 5 | 590 | |||
DA032832.1 | - | 3 | 300 | |||
DA160782.1 | - | 4 | 452 | |||
DA386385.1 | - | 3 | 282 | |||
DA704994.1 | - | 4 | 420 |