Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297760.1 | NP_001284689.1 | 2 | 782 | ||
NM_001297762.1 | NP_001284691.1 | 2 | 782 | |||
NM_018208.3 | NP_060678.2 | 2 | 782 | |||
GenBank mRNA | AB073608.1 | - | 2 | 382 | ||
AK001623.1 | - | 2 | 494 | |||
BC010082.2 | - | 2 | 603 | 4329734 |