Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018225.2 | NP_060695.2 | 10 | 1349 | ||
XM_005251503.4 | XP_005251560.1 | 9 | 1185 | |||
GenBank mRNA | AK001667.1 | - | 10 | 1279 | ||
AK022032.1 | - | 2 | 153 | |||
AK225559.1 | - | 10 | 1304 | |||
AK225587.1 | - | 10 | 1298 | |||
AK304767.1 | - | 8 | 936 | |||
BC002876.2 | - | 10 | 1295 | 3940669 | ||
BC110654.1 | - | 10 | 1389 | 6168993 | ||
EF011613.1 | - | 10 | 1295 |