Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018268.3 | NP_060738.2 | 12 | 1457 | ||
XM_005248551.4 | XP_005248608.1 | 10 | 1250 | |||
XM_005248552.3 | XP_005248609.1 | 12 | 1425 | |||
XM_011543505.2 | XP_011541807.1 | 12 | 1219 | |||
XM_011543507.2 | XP_011541809.1 | 11 | 1335 | |||
XM_017009606.1 | XP_016865095.1 | 12 | 1867 | |||
GenBank mRNA | AF115511.1 | - | 12 | 1205 | ||
AK001766.1 | - | 12 | 1403 | |||
AK021855.1 | - | 12 | 1243 | |||
AK023977.1 | - | 12 | 1159 | |||
AK293316.1 | - | 10 | 942 | |||
AK300058.1 | - | 10 | 1068 | |||
AK304329.1 | - | 12 | 1149 | |||
AL834138.1 | - | 6 | 2138 | |||
BC040241.1 | - | 12 | 1209 | 3447269 | ||
KU178690.1 | - | 12 | 1137 | |||
KU178691.1 | - | 12 | 1152 |