Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CASC1 SIRNA Assays ›
Gene Symbol:
CASC1
Gene Name:
cancer susceptibility candidate 1
Gene Aliases:
LAS1, PPP1R54
Chromosome Location:
Chr.12: 25107057 - 25195177 on Build GRCh38
UniGene ID:
Hs.407771
Species:
Human
siRNA ID
HSS182980
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CASC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001082972.2
NP_001076441.1
5
547
NM_001082973.2
NP_001076442.1
4
339
NM_001204101.2
NP_001191030.1
2
162
NM_001204102.2
NP_001191031.1
6
395
NM_001319977.1
NP_001306906.1
3
247
NM_001319978.1
NP_001306907.1
4
513
NM_018272.4
NP_060742.3
5
357
XM_005253406.1
XP_005253463.1
5
471
XM_005253408.1
XP_005253465.1
4
233
XM_005253409.1
XP_005253466.1
3
247
XM_006719101.1
XP_006719164.1
5
471
XM_011520723.1
XP_011519025.1
5
471
XM_011520726.1
XP_011519028.1
5
471
XM_011520727.1
XP_011519029.1
5
471
XM_017019560.1
XP_016875049.1
4
436
XM_017019561.1
XP_016875050.1
3
443
XM_017019562.1
XP_016875051.1
4
321
XM_017019563.1
XP_016875052.1
5
353
XM_017019564.1
XP_016875053.1
4
321
XM_017019565.1
XP_016875054.1
5
471
XM_017019566.1
XP_016875055.1
5
682
XM_017019567.1
XP_016875056.1
2
126
XM_017019568.1
XP_016875057.1
3
265
GenBank mRNA
AK093102.1
-
5
325
AK297998.1
-
2
126
AK301763.1
-
5
358
AK302235.1
-
4
303
AK304602.1
-
3
247
AY423543.1
-
3
255
BC028415.1
-
5
488
4823620
BC031047.1
-
5
339
BC047415.1
-
6
393
5269578
BC057795.1
-
4
337
5266518
BC117281.1
-
2
172
BC117283.1
-
2
172
DA753279.1
-
5
385
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-kq4fm:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline