Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039702.2 | NP_001034791.1 | 8 | 986 | ||
NM_018324.2 | NP_060794.1 | 9 | 1145 | |||
XM_017016376.1 | XP_016871865.1 | 9 | 1109 | |||
GenBank mRNA | AK001844.1 | - | 8 | 936 | ||
AK001968.1 | - | 9 | 1078 | |||
BC050372.1 | - | 8 | 953 | 5742377 | ||
BG281536.1 | - | 2 | 171 |