Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018346.2 | NP_060816.1 | 4 | 846 | ||
NR_130911.1 | 2 | 507 | ||||
GenBank mRNA | AK002026.1 | - | 4 | 817 | ||
AK022105.1 | - | 4 | 786 | |||
AK222472.1 | - | 4 | 797 | |||
AK297655.1 | - | 2 | 447 | |||
BC005854.2 | - | 3 | 598 | |||
BC050538.1 | - | 4 | 777 | 5214885 |