Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018392.4 | NP_060862.3 | 5 | 399 | ||
XM_005263115.3 | XP_005263172.1 | 5 | 454 | |||
XM_011532091.2 | XP_011530393.1 | 5 | 399 | |||
XM_011532092.2 | XP_011530394.1 | 4 | 279 | |||
XM_011532093.2 | XP_011530395.1 | 5 | 399 | |||
XM_011532094.2 | XP_011530396.1 | 5 | 399 | |||
XM_011532096.2 | XP_011530398.1 | 5 | 399 | |||
XM_017008369.1 | XP_016863858.1 | 4 | 394 | |||
XM_017008370.1 | XP_016863859.1 | 5 | 399 | |||
XM_017008371.1 | XP_016863860.1 | 4 | 279 | |||
XM_017008372.1 | XP_016863861.1 | 5 | 308 | |||
XM_017008373.1 | XP_016863862.1 | 4 | 245 | |||
GenBank mRNA | AK002193.1 | - | 5 | 316 | ||
AK299831.1 | - | 5 | 399 | |||
AK302613.1 | - | 6 | 624 | |||
AL708767.1 | - | 5 | 322 | |||
BC044799.1 | - | 5 | 304 | 4836807 |